2,4,6-Trimethoxyanilin Hydrochlorid

Lieferant: BIONET RESEARCH

8W-0406S.1 8W-0406S.10 8W-0406S.5 8W-0406S.500
BIOT8W-0406S.1EA 0 EUR
BIOT8W-0406S.1 BIOT8W-0406S.10 BIOT8W-0406S.5 BIOT8W-0406S.500
2,4,6-Trimethoxyanilin Hydrochlorid
2,4,6-Trimethoxyanilin Hydrochlorid
Formel: C₉H₁₄ClNO₃
Molecular Weight: 219,665 g/mol
MDL: MFCD00211326
CAS-Nummer: 102438-99-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR