(S)-(+)-1-(1-Naphthyl)ethylisocyanat 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(S)-(+)-1-(1-Naphthyl)ethylisocyanate
L20242.03
L20242.MD
L20242.03EA
90.8
EUR
L20242.03
L20242.MD
(S)-(+)-1-(1-Naphthyl)ethylisocyanat 95%
(S)-(+)-1-(1-Naphthyl)ethylisocyanat
Purity: 95%
CAS No.: 73671-79-1
Molecular Formula: C13H11NO
Formula Weight: 197.24
Storage Temperature: 2°C to 8°C
Flash Point: 93°(199°F)
Refractive Index: 1.6045
Boiling Point: 106-108°/0.16m
Beilstein Reference: 4675069
Optical Rotation: +47° (c=3.5 in toluene)
UN No.: 2206
MDL No.: MFCD00064181
Formel:
C₁₃H₁₁NO Molecular Weight: 197,24 g/mol Siedepunkt: 106…108 °C (0,16 m) Dichte: 1,118 Flammpunkt: 93 °C (199 °F) |
MDL:
MFCD00064181 CAS-Nummer: 73671-79-1 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...