4-Chlorbenzoylisothiocyanat

Lieferant: FLUOROCHEM

002526-5G 002526-25G
FLUO002526-5GEA 0 EUR
FLUO002526-5G FLUO002526-25G
4-Chlorbenzoylisothiocyanat
4-Chlorbenzoylisothiocyanat
Formel: C₈H₄ClNOS
MDL: MFCD00041104
CAS-Nummer: 16794-67-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR