4-(Trifluormethoxy)benzylmercaptan
Lieferant: FLUOROCHEM
Synonyme:
4-(Trifluoromethoxy)benzylthiol
007554-1G
007554-5G
FLUO007554-1GEA
122
EUR
FLUO007554-1G
FLUO007554-5G
4-(Trifluormethoxy)benzylmercaptan
4-(Trifluormethoxy)benzylmercaptan
Formel:
C₈H₇F₃OS |
MDL:
MFCD00052327 CAS-Nummer: 175278-03-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...