4-(Trifluormethoxy)benzylmercaptan

Lieferant: Thermo Scientific

Synonyme: 4-(Trifluoromethoxy)benzylthiol

TL00388.250 TL00388DA
MAYBTL00388.250EA 0 EUR
MAYBTL00388.250 MAYBTL00388DA
4-(Trifluormethoxy)benzylmercaptan
4-(Trifluormethoxy)benzylmercaptan
Formel: C₈H₇F₃OS
MDL: MFCD00052327
CAS-Nummer: 175278-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR