4,4'-(Hexafluorisopropyliden)diphenol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4,4′-(Hexafluoroisopropylidene)diphenol, 2,2-Bis(4-hydroxyphenyl)hexafluoropropane, 4,4'-(Perfluoropropane-2,2-diyl)diphenol
M-Clarity™ Qualitätsstufe = MQ100
257591-100G
257591-25G
SIAL257591-100GEA
778
EUR
SIAL257591-100G
SIAL257591-25G
4,4'-(Hexafluorisopropyliden)diphenol, Sigma-Aldrich®
4,4'-(Hexafluorisopropyliden)diphenol
Formel:
C₁₅H₁₀F₆O₂ Molecular Weight: 336.24 g/mol |
MDL:
MFCD00000439 CAS-Nummer: 1478-61-1 EINECS: 216-036-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...