2-Biphenylylisocyanat

Lieferant: Thermo Scientific

Synonyme: 2-Biphenyl Isocyanate, Isocyanic acid 2-Biphenyl ester, 2-Biphenylylisocyanate

AC31411DA AC31411EA AC31411.50
MAYBAC31411DAEA 0 EUR
MAYBAC31411DA MAYBAC31411EA MAYBAC31411.50
2-Biphenylylisocyanat
2-Biphenylylisocyanat
Formel: C₁₃H₉NO
Molecular Weight: 195,2167 g/mol
MDL: MFCD00037087
CAS-Nummer: 17337-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR