Ethyl-2-oxocyclohexancarboxylat 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 2-oxocyclohexanecarboxylate

118330050 118330250 118331000
ACRO118330050EA 19.7 EUR
ACRO118330050 ACRO118330250 ACRO118331000
Ethyl-2-oxocyclohexancarboxylat 95%
Ethyl-2-oxocyclohexancarboxylat
Formel: C₉H₁₄O₃
Molecular Weight: 170,21 g/mol
Siedepunkt: 106 °C (11 mmHg)
Dichte: 1,06 g/cm³
Flammpunkt: 85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001631
CAS-Nummer: 1655-07-8
EINECS: 216-731-5

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4775 to 1.4795 (20°C, 589 nm)
GC ≥94 %
Molybdenum (Mo) ≤10 ppm
Heavy metals & Fe (as Pb) ≤0.001 %
Titrable free base ≤0.02 meq/g
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR