Ethyl-2-oxocyclohexancarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 2-oxocyclohexanecarboxylate

M-Clarity™ Qualitätsstufe = MQ100

166995-100G 166995-25G
SIAL166995-100GEA 329 EUR
SIAL166995-100G SIAL166995-25G
Ethyl-2-oxocyclohexancarboxylat, Sigma-Aldrich®
Ethyl-2-oxocyclohexancarboxylat
Formel: C₉H₁₄O₃
Molecular Weight: 170,21 g/mol
MDL: MFCD00001631
CAS-Nummer: 1655-07-8
EINECS: 216-731-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR