Ethyl-2-oxocyclohexancarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 2-oxocyclohexanecarboxylate

M-Clarity™ Qualitätsstufe = MQ200

E20004-100G E20004-25G
SIALE20004-100GEA 257 EUR
SIALE20004-100G SIALE20004-25G
Ethyl-2-oxocyclohexancarboxylat, Sigma-Aldrich®
Ethyl-2-oxocyclohexancarboxylat
Formel: C₉H₁₄O₃
Molecular Weight: 170,21 g/mol
MDL: MFCD00001631
CAS-Nummer: 1655-07-8
EINECS: 216-731-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR