Methyl-2-oxocyclohexancarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2-oxocyclohexanecarboxylate

29162-10ML 29162-50ML
SIAL29162-10MLEA 284 EUR
SIAL29162-10ML SIAL29162-50ML
Methyl-2-oxocyclohexancarboxylat, Sigma-Aldrich®
Methyl-2-oxocyclohexancarboxylat
Formel: C₈H₁₂O₃
Molecular Weight: 156,1791 g/mol
MDL: MFCD00077699
CAS-Nummer: 41302-34-5
EINECS: 255-306-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR