Ethyl-3-oxocyclohexancarboxylat
Lieferant: CHEMPUR
Synonyme:
Ethyl 3-oxocyclohexane-1-carboxylate
RJC00222.250
RJC00222.1
CHMPRJC00222.250EA
0
EUR
CHMPRJC00222.250
CHMPRJC00222.1
Ethyl-3-oxocyclohexancarboxylat
Ethyl-3-oxocyclohexancarboxylat
Formel:
C₉H₁₄O₃ Molecular Weight: 170,2100 g/mol |
MDL:
MFCD00205583 CAS-Nummer: 33668-25-6 EINECS: 251-626-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...