4-(tert-Butyl)cyclohexanonoxim

Lieferant: BIONET RESEARCH

Synonyme: N-(4-tert-Butylcyclohexylidene)hydroxylamine

3T-1121.500 3T-1121.1 3T-1121.5 3T-1121.10
BIOT3T-1121.500EA 0 EUR
BIOT3T-1121.500 BIOT3T-1121.1 BIOT3T-1121.5 BIOT3T-1121.10
4-(tert-Butyl)cyclohexanonoxim
4-(tert-Butyl)cyclohexanonoxim
Formel: C₁₀H₁₉NO
CAS-Nummer: 4701-98-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR