O-tert-Butylhydroxylamin Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme: O-(tert-Butyl)hydroxylamine hydrochloride
20023-1G 20023-5G
SIAL20023-1GEA 64.3 EUR
SIAL20023-1G SIAL20023-5G
O-tert-Butylhydroxylamin Hydrochlorid, Sigma-Aldrich®
O-tert-Butylhydroxylamin Hydrochlorid
Formel: C₄H₁₂ClNO
Molecular Weight: 125,5972 g/mol
MDL: MFCD00043272
CAS-Nummer: 39684-28-1
EINECS: 254-590-1