O-Methylhydroxylamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methylhydroxylamine hydrochloride, O-Methylhydroxylammonium chloride, Methoxyamine hydrochloride, Methoxylamine hydrochloride

M-Clarity™ Qualitätsstufe = MQ200

226904-5G 226904-100G 226904-1G 226904-25G
SIAL226904-5GEA 61.2 EUR
SIAL226904-5G SIAL226904-100G SIAL226904-1G SIAL226904-25G
O-Methylhydroxylamin Hydrochlorid, Sigma-Aldrich®
O-Methylhydroxylamin Hydrochlorid
Formel: CH₆ClNO
Molecular Weight: 83,515 g/mol
MDL: MFCD00012951
CAS-Nummer: 593-56-6
EINECS: 209-798-7
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR