Diethylchlorphosphit 90% tech.
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
114250250
114251000
ACRO114250250EA
105
EUR
ACRO114250250
ACRO114251000
Diethylchlorphosphit 90% tech.
Diethylchlorphosphit
Formel:
C₄H₁₀ClO₂P Molecular Weight: 156.55 g/mol Dichte: 1.09 g/cm³ Flammpunkt: <1 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009074 CAS-Nummer: 589-57-1 EINECS: 209-652-2 UN: 2924 ADR: 3 II |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4340 to 1.4410 (20°C, 589 nm) |
Titration Argentometric | ≥88 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...