Mono(trimethylsilyl)phosphit
Lieferant: CHEMPUR SILANE
SILS20020.1
SILS20020.5
SILS20020.25
SILNSILS20020.1EA
0
EUR
SILNSILS20020.1
SILNSILS20020.5
SILNSILS20020.25
Mono(trimethylsilyl)phosphit
Mono(trimethylsilyl)phosphit
Formel:
C₃H₁₁O₃PSi |
MDL:
MFCD00800498 CAS-Nummer: 91076-68-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...