Tris(trimethylsilyl)phosphit, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tris(trimethylsilyl)phosphite

93412-100ML 93412-25ML 93412-5ML
SIAL93412-100MLEA 990 EUR
SIAL93412-100ML SIAL93412-25ML SIAL93412-5ML
Tris(trimethylsilyl)phosphit, Sigma-Aldrich®
Tris(trimethylsilyl)phosphit
Formel: C₉H₂₇O₃PSi₃
Molecular Weight: 298,54 g/mol
MDL: MFCD00015588
CAS-Nummer: 1795-31-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR