Tetrakis(diethylamino)titan (TDEAT), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetrakis(diethylamino)titanium(IV)

M-Clarity™ Qualitätsstufe = MQ100

469866-25G 469866-5G
SIAL469866-25GEA 746 EUR
SIAL469866-25G SIAL469866-5G
Tetrakis(diethylamino)titan (TDEAT), Sigma-Aldrich®
Tetrakis(diethylamino)titan (TDEAT)
Formel: C₁₆H₄₀N₄Ti
Molecular Weight: 336,383 g/mol
MDL: MFCD00015648
CAS-Nummer: 4419-47-0
EINECS: 437-500-7
UN: 3129
ADR: 4.3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR