Tetrakis(dimethylamido)titan ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Tetrakis(dimethylamido)titan(IV), Tetrakis(dimethylamino)titan(IV), TDMAT, Tetrakis(dimethylamino)titan
42932.06
42932.14
42932.06EA
213
EUR
42932.06
42932.14
Tetrakis(dimethylamido)titan ≥99,9% (Metall-Basis)
Tetrakis(dimethylamido)titan
Purity: 99.9%
CAS No.: 3275-24-9
Molecular Formula: C8H24N4Ti
Formula Weight: 224.19
Flash Point: -30°(-22°F)
Physical Form: Liquid
Density: 0.96
Sensitivity: Moisture Sensitive
Appearance: Yellow-brown
Boiling Point: 50°/0.5mm
UN No.: 3129
MDL No.: MFCD00014861
Formel:
C₈H₂₄N₄Ti Molecular Weight: 224.17 g/mol Siedepunkt: 50 °C (0.5 mmHg) Dichte: 0.96 Flammpunkt: –30 °C (–22 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014861 CAS-Nummer: 3275-24-9 EINECS: 221-904-3 UN: 3129 ADR: 4.3 II |