Tetrakis(dimethylamido)titan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Tetrakis(dimethylamido)titan(IV), Tetrakis(dimethylamino)titan(IV), TDMAT, Tetrakis(dimethylamino)titan
M-Clarity™ Qualitätsstufe = MQ200
469858-25G
469858-5G
SIAL469858-25GEA
884
EUR
SIAL469858-25G
SIAL469858-5G
Tetrakis(dimethylamido)titan, Sigma-Aldrich®
Tetrakis(dimethylamido)titan
Formel:
C₈H₂₄N₄Ti Molecular Weight: 224,17 g/mol |
MDL:
MFCD00014861 CAS-Nummer: 3275-24-9 EINECS: 221-904-3 UN: 3129 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...