2-(1,1,2,2-Tetrafluorethoxy)benzoesäure

Lieferant: FLUOROCHEM

Synonyme: 2-(2H-Perfluoroethoxy)benzoic acid

010531-25G 010531-5G
FLUO010531-25GEA 0 EUR
FLUO010531-25G FLUO010531-5G
2-(1,1,2,2-Tetrafluorethoxy)benzoesäure
2-(1,1,2,2-Tetrafluoroethoxy)benzoic acid
Formel: C₉H₆F₄O₃
MDL: MFCD03094352
CAS-Nummer: 10008-97-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR