3-(1,1,2,2-Tetrafluorethoxy)benzoesäure

Lieferant: CHEMPUR

FL9793.POR 009793-1G 009793-5G
CHMPFL9793.POREA 0 EUR
CHMPFL9793.POR FLUO009793-1G FLUO009793-5G
3-(1,1,2,2-Tetrafluorethoxy)benzoesäure
3-(1,1,2,2-Tetrafluoroethoxy)benzoic acid
Formel: C₉H₆F₄O₃
MDL: MFCD00155925
CAS-Nummer: 70126-48-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR