4-(1,1,2,2-Tetrafluorethoxy)benzoesäure

Lieferant: FLUOROCHEM

010533-25G 010533-5G
FLUO010533-25GEA 1040 EUR
FLUO010533-25G FLUO010533-5G
4-(1,1,2,2-Tetrafluorethoxy)benzoesäure
4-(1,1,2,2-Tetrafluoroethoxy)benzoic acid
Formel: C₉H₆F₄O₃
MDL: MFCD00155926
CAS-Nummer: 10009-25-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR