3-Hydroxypyridin-N-oxid

Lieferant: CHEMPUR

Synonyme: 3-Pyridinol N-Oxide, 3-Hydroxypyridine 1-oxide

BTB04151.250 BTB04151.1
CHMPBTB04151.250EA 0 EUR
CHMPBTB04151.250 MAYBBTB04151.1
3-Hydroxypyridin-N-oxid
3-Hydroxypyridin-N-oxid
Formel: C₅H₅NO₂
Molecular Weight: 111,0987 g/mol
MDL: MFCD00006200
CAS-Nummer: 6602-28-4
EINECS: 229-545-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR