2-Mercaptopyridin-N-oxid Zinksalz, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(2-pyridylthio) Zinc 1,1'-Dioxide, Zinc Pyrithione, 2-Pyridinethiol 1-Oxide Zinc salt, 1-Hydroxypyridine-2-thione Zinc salt

H6377-100G H6377-10G
SIALH6377-100GEA 274 EUR
SIALH6377-100G SIALH6377-10G
2-Mercaptopyridin-N-oxid Zinksalz, Sigma-Aldrich®
2-Mercaptopyridin-N-oxid Zinksalz
Formel: C₁₀H₈N₂O₂S₂Zn
Molecular Weight: 317.72 g/mol
MDL: MFCD00067336
CAS-Nummer: 13463-41-7
EINECS: 236-671-3
UN: 2811
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR