4-Benzyloxy-2-hydroxypyridin ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Benzyloxy-2(1H)-pyridone
B24774.06
B24774.14
B24774.06EA
103
EUR
B24774.06
B24774.14
4-Benzyloxy-2-hydroxypyridin ≥97%
4-Benzyloxy-2-hydroxypyridin
Purity: 97%
CAS No.: 53937-02-3
Molecular Formula: C12H11NO2
Formula Weight: 201.22
Melting Point: 194-198°
MDL No.: MFCD04039771
Formel:
C₁₂H₁₁NO₂ Molecular Weight: 201,22 g/mol Schmelzpunkt: 194…198 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191783 CAS-Nummer: 53937-02-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...