5-Acetyl-2-thiophenboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Acetylthien-5-yl)boronic acid,(5-Acetylthien-2-yl)boronic acid,5-Acetyl-2-thiopheneboronic acid,5-Acetylthiophen-2-ylboronic acid, 5-Acetyl-2-thienylboronic acid, 5-Acetylthiophene-2-boronic acid, 5-Acetyl-2-thiopheneboronic acid

BB-2011-1G BB-2011-5G BB-2011-25G
COBBBB-2011-1GEA 26.2 EUR
COBBBB-2011-1G COBBBB-2011-5G COBBBB-2011-25G
5-Acetyl-2-thiophenboronsäure
5-Acetyl-2-thiophenboronsäure
Formel: C₆H₇BO₃S
Molecular Weight: 170,00 g/mol
MDL: MFCD01075681
CAS-Nummer: 206551-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR