2-Acetyl-5-cyanthiophen

Lieferant: FLUOROCHEM

Synonyme: 5-Acetylthiophene-2-carbonitrile

CFF191.2.5 CFF191.10 CFF191.25 017260-2.5G 017260-10G 017260-25G
CHMPCFF191.2.5EA 268 EUR
CHMPCFF191.2.5 CHMPCFF191.10 CHMPCFF191.25 FLUO017260-2.5G FLUO017260-10G FLUO017260-25G
2-Acetyl-5-cyanthiophen
2-Acetyl-5-cyanthiophen
Formel: C₇H₅NOS
Molecular Weight: 151.19 g/mol
MDL: MFCD00052314
CAS-Nummer: 88653-55-8
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR