2-Acetyl-5-cyanthiophen

Lieferant: Thermo Scientific

Synonyme: 5-Acetylthiophene-2-carbonitrile

TL00354.1 TL00354EA
MAYBTL00354.1EA 0 EUR
MAYBTL00354.1 MAYBTL00354EA
2-Acetyl-5-cyanthiophen
2-Acetyl-5-cyanthiophen
Formel: C₇H₅NOS
Molecular Weight: 151.19 g/mol
MDL: MFCD00052314
CAS-Nummer: 88653-55-8
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR