5-Acetyl-2-thiophencarbonsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Acetylthiophene-2-carboxylic Acid, 5-Acetyl-2-thenoic acid
L13588.06
L13588.14
L13588.06EA
63.8
EUR
L13588.06
L13588.14
5-Acetyl-2-thiophencarbonsäure ≥98%
5-Acetyl-2-thiophencarbonsäure
Purity: 98+%
CAS No.: 4066-41-5
Molecular Formula: C7H6O3S
Formula Weight: 170.19
Melting Point: 208-212°
Beilstein Reference: 128897
Formel:
C₇H₆O₃S Molecular Weight: 170,19 g/mol Schmelzpunkt: 208…212 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00055512 CAS-Nummer: 4066-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...