2-(2,2,2-Trifluoracetyl)thiophen ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-(Trifluoroacetyl)thiophene
L12585.03
L12585.06
L12585.03EA
40.4
EUR
L12585.03
L12585.06
2-(2,2,2-Trifluoracetyl)thiophen ≥98%
2-(2,2,2-Trifluoracetyl)thiophen
Purity: 98%
CAS No.: 651-70-7
Molecular Formula: C6H3F3OS
Formula Weight: 180.15
Flash Point: 58°(136°F)
Refractive Index: 1.4845
Boiling Point: 163-165°
Beilstein Reference: 1309208
UN No.: 1993
MDL No.: MFCD00041427
Formel:
C₆H₅ClOS Molecular Weight: 180,15 g/mol Siedepunkt: 163…165 °C Dichte: 1,403 Flammpunkt: 58 °C (136 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041427 CAS-Nummer: 651-70-7 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...