2-Acetyl-3-amino-5-tert-butylthiophen

Lieferant: FLUOROCHEM

CFF500.5 CFF500.25 017508-5G 017508-25G
CHMPCFF500.5EA 348 EUR
CHMPCFF500.5 CHMPCFF500.25 FLUO017508-5G FLUO017508-25G
2-Acetyl-3-amino-5-tert-butylthiophen
2-Acetyl-3-amino-5-tert-butylthiophen
Formel: C₁₀H₁₅NOS
MDL: MFCD00068131
CAS-Nummer: 175137-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR