Schwefeltrioxid-Trimethylamin-Komplex 95%
Lieferant: Thermo Scientific
|
Gefahr
|
L00302.14
L00302.22
L00302.36
L00302.14EA
37.3
EUR
L00302.14
L00302.22
L00302.36
Schwefeltrioxid-Trimethylamin-Komplex 95%
Schwefeltrioxid-Trimethylamin-Komplex
Purity: 95%
CAS No.: 3162-58-1
Molecular Formula: C3H9NO3S
Formula Weight: 139.17
Melting Point: ca 235° dec.
Beilstein Reference: 3681759
UN No.: 3261
MDL No.: MFCD00012421
Formel:
C₃H₉NO₃S Molecular Weight: 139,17 g/mol Schmelzpunkt: ∼ 235 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012421 CAS-Nummer: 3162-58-1 EINECS: 221-614-7 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...