Methylmethacrylat-[D8] (monomer) (99% D), Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: Methyl methacrylate-[²H₈] (monomer)

M-Clarity™ Qualitätsstufe = MQ200

444960-1G 444960-5G
SIAL444960-1GEA 319 EUR
SIAL444960-1G SIAL444960-5G
Methylmethacrylat-[D8] (monomer) (99% D), Sigma-Aldrich®
Methylmethacrylat-[D8] (monomer)
Formel: C₅D₈O₂
CAS-Nummer: 35233-69-3
EINECS: 252-449-9
UN: 1247
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR