(±)-20-HDoHE
Lieferant: Cayman Chemical
33750-25
33750-50
33750-100
33750-250
CAYM33750-25EA
103
EUR
CAYM33750-25
CAYM33750-50
CAYM33750-100
CAYM33750-250
(±)-20-HDoHE
(±)-20-HDoHE
Formel:
C₂₂H₃₂O₃ |
CAS-Nummer:
90906-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...