(±)-16-HDoHE

Lieferant: Cayman Chemical

33600-25 33600-50 33600-100 33600-250
CAYM33600-25EA 103 EUR
CAYM33600-25 CAYM33600-50 CAYM33600-100 CAYM33600-250
(±)-16-HDoHE
(±)-16-HDoHE
Formel: C₂₂H₃₂O₃
CAS-Nummer: 90780-51-1
UN: 1170
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR