(±)-8-HDoHE

Lieferant: Cayman Chemical

33350-25 33350-50 33350-100 33350-250
CAYM33350-25EA 103 EUR
CAYM33350-25 CAYM33350-50 CAYM33350-100 CAYM33350-250
(±)-8-HDoHE
(±)-8-HDoHE
Formel: C₂₂H₃₂O₃
CAS-Nummer: 90780-54-4
UN: 1170
ADR: 3 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR