(R)-17-HDoHE

Lieferant: Cayman Chemical

10005099-25 10005099-50 10005099-100 10005099-250
CAYM10005099-25EA 161 EUR
CAYM10005099-25 CAYM10005099-50 CAYM10005099-100 CAYM10005099-250
(R)-17-HDoHE
(R)-17-HDoHE
Formel: C₂₂H₃₂O₃
CAS-Nummer: 155976-53-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR