(R)-(+)-α,α-Diphenylprolinol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(R)-(+)-alpha,alpha-Diphenylprolinol, (R)-(+)-α,α-Diphenyl-2-pyrrolidinemethanol, (R)-(+)-2-(Diphenylhydroxymethyl)pyrrolidine, α,α-Diphenyl-D-prolinol
382337-100MG
382337-1G
382337-5G
SIAL382337-100MGEA
118
EUR
SIAL382337-100MG
SIAL382337-1G
SIAL382337-5G
(R)-(+)-α,α-Diphenylprolinol, Sigma-Aldrich®
(R)-(+)-α,α-Diphenylprolinol
Formel:
C₁₇H₁₉NO Molecular Weight: 253,35 g/mol |
MDL:
MFCD00077754 CAS-Nummer: 22348-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...