(S,S)-(-)-2,3-Bis(diphenylphosphino)butan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2S,3S)-(-)-Bis(diphenylphosphino)butane, (2S,3S)-Butane-2,3-diylbis(diphenylphosphine), (S,S)-CHIRAPHOS

M-Clarity™ Qualitätsstufe = MQ100

259098-1G 259098-250MG
SIAL259098-1GEA 637 EUR
SIAL259098-1G SIAL259098-250MG
(S,S)-(-)-2,3-Bis(diphenylphosphino)butan, Sigma-Aldrich®
(S,S)-(-)-2,3-Bis(diphenylphosphino)butan
Formel: C₂₈H₂₈P₂
Molecular Weight: 426,47 g/mol
MDL: MFCD00009836
CAS-Nummer: 64896-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR