Ethyl-2-(5-brom-2-thienyl)-2-oxoacetat

Lieferant: Thermo Scientific

BTB09133DA BTB09133EA
MAYBBTB09133DAEA 0 EUR
MAYBBTB09133DA MAYBBTB09133EA
Ethyl-2-(5-brom-2-thienyl)-2-oxoacetat
Ethyl-2-(5-brom-2-thienyl)-2-oxoacetat
Formel: C₈H₇BrO₃S
CAS-Nummer: 22098-10-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR