Hydrindantin Dihydrat, Supelco®

Lieferant: Merck

Synonyme: (2,2'-Biindan)-1,1',3,3'-tetrone, 2,2'-dihydroxy-, 1,1'-dihydrate (7CI), 2,2',3,3,3',3'-Hexahydroxy-2,2'-biindan-1,1'-dione

M-Clarity™ Qualitätsstufe = MQ100

53940-10G 53940-50G
53940-10GEA 112 EUR
53940-10G 53940-50G
Hydrindantin Dihydrat, Supelco®
Hydrindantin Dihydrat
Formel: C₁₈H₁₀O₆·2H₂O
Molecular Weight: 358,30 g/mol
MDL: MFCD00149242
CAS-Nummer: 5950-69-6
EINECS: 227-713-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fd0cbf04561c981223857aa80e2e48f97a4d548&vid=9c1e9f587130e5ce2cc51e48cdbcec3800d035a1&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR