Hydrindantin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

H17309-25G H17309-5G
SIALH17309-25GEA 453 EUR
SIALH17309-25G SIALH17309-5G
Hydrindantin, Sigma-Aldrich®
Hydrindantin
Formel: C₁₈H₁₀O₆
CAS-Nummer: 5103-42-4
EINECS: 225-823-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR