2-(Hydroxymethyl)-18-krone-6 ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 97%
CAS No.: 70069-04-4
Molecular Formula: C13H27O7
Formula Weight: 294.35
Flash Point:
Formel:
C₁₃H₂₆O₇ Siedepunkt: 150 °C (0,5 mmHg) Dichte: 1,174 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00188046 CAS-Nummer: 70069-04-4 EINECS: 274-299-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...