2-Chlor-6-(1-imidazolyl)pyrazin

Lieferant: COMBI-BLOCKS

HC-6148-5G HC-6148-1G
COBBHC-6148-5GEA 595 EUR
COBBHC-6148-5G COBBHC-6148-1G
2-Chlor-6-(1-imidazolyl)pyrazin
2-Chloro-6-(1-imidazolyl)pyrazine
Formel: C₇H₅ClN₄
MDL: MFCD09800962
CAS-Nummer: 941294-48-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR