1-(2-Tetrahydropyranyl)-1H-pyrazol-5-boronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-(Tetrahydropyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole-5-boronic acid Pinacol ester, 1-(Tetrahydropyran-2-yl)-1H-pyrazole-5-boronic acid pinacol ester, 1-(Tetrahydro-2H-pyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
718823-1G
718823-5G
SIAL718823-1GEA
39.6
EUR
SIAL718823-1G
SIAL718823-5G
1-(2-Tetrahydropyranyl)-1H-pyrazol-5-boronsäurepinakolester, Sigma-Aldrich®
1-(2-Tetrahydropyranyl)-1H-pyrazol-5-boronsäurepinakolester
Formel:
C₁₄H₂₃BN₂O₃ Molecular Weight: 278,16 g/mol |
MDL:
MFCD09037501 CAS-Nummer: 903550-26-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...