(2-Nitrophenyl)essigsäure

Lieferant: Thermo Scientific

Synonyme: 2-(2-Nitrophenyl)acetic acid

SB01683.1 SB01683.10 SB01683.50
MAYBSB01683.1EA 0 EUR
MAYBSB01683.1 MAYBSB01683.10 MAYBSB01683.50
(2-Nitrophenyl)essigsäure
(2-Nitrophenyl)essigsäure
Formel: (C₆H₄NO₂)CH₂COOH
Molecular Weight: 181.15 g/mol
Schmelzpunkt: 136…139 °C
MDL: MFCD00007190
CAS-Nummer: 3740-52-1
EINECS: 223-128-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR