(2-Iodphenyl)essigsäure

Lieferant: CHEMPUR
Achtung

Synonyme: 2-(2-Iodophenyl)acetic acid

NRB00709.250 NRB00709.1
CHMPNRB00709.250EA 0 EUR
CHMPNRB00709.250 MAYBNRB00709.1
(2-Iodphenyl)essigsäure
(2-Iodphenyl)essigsäure
Formel: (C₆H₄I)CH₂COOH
Molecular Weight: 262,05 g/mol
MDL: MFCD00046546
CAS-Nummer: 18698-96-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR