Ammoniumperoxodisulfat (APS) analytisches Reagens

Lieferant: Bernd Kraft
Gefahr

Synonyme: Peroxidisulphuric acid diammonium salt, Ammonium persulphate

20784.0600 20784.2600 20784.3600 20784.5600
KRAF20784.0600EA 28.5 EUR
KRAF20784.0600 KRAF20784.2600 KRAF20784.3600 KRAF20784.5600
Ammoniumperoxodisulfat (APS) analytisches Reagens
Ammoniumperoxodisulfat (APS)
Formel: (NH₄)₂S₂O₈
Molecular Weight: 228.2 g/mol
Schmelzpunkt: 120
Dichte: 1.98 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003390
CAS-Nummer: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1 III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR